3-Fluorocyclobutanamine hydrochloride 97%

Supplier: Ambeed
Warning

A746530-1G A746530-250MG A746530-100MG
77171-016EA 253.38 USD
77171-016 77171-018 77614-292
3-Fluorocyclobutanamine hydrochloride 97%
3-Fluorocyclobutanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₈FN·ClH
MW: 125.572 g/mol
MDL Number: MFCD21642364
CAS Number: 1284245-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR