1,1-Dimethylurea 96%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethylurea

A746501-100G A746501-1G A746501-250MG A746501-25G A746501-5G
77199-186EA 156.56 USD
77199-186 77199-188 77199-190 77199-192 77199-194
1,1-Dimethylurea 96%
1,1-Dimethylurea

Precaución: for Research Use Only

Formula: C₃H₈N₂O
MW: 88.11 g/mol
MDL Number: MFCD00007959
CAS Number: 598-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR