Diethyl (bromomethyl)phosphonate 98%

Supplier: Ambeed
Warning

A746319-1G A746319-250MG A746319-5G
77171-010EA 116.39 USD
77171-010 77171-012 77171-014
Diethyl (bromomethyl)phosphonate 98%
Diethyl (bromomethyl)phosphonate

Precaución: for Research Use Only

MDL Number: MFCD00972940
CAS Number: 66197-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR