Dichloro(N,N,N',N'-tetramethylethylenediamine)palladium(II) 98% Pd> 36.2%

Supplier: Ambeed
Warning

A746271-100MG A746271-1G A746271-250MG
77171-004EA 13 USD
77171-004 77171-006 77171-008
Dichloro(N,N,N',N'-tetramethylethylenediamine)palladium(II) 98% Pd> 36.2%
Dichloro(N,N,N',N'-tetramethylethylenediamine)palladium(II)

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂Pd
MDL Number: MFCD00075317
CAS Number: 14267-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR