Leucoquinizarin 96%

Supplier: Ambeed
Warning

Synonyms: Anthracene-1,4,9,10-tetraol, leuco-Quinizarin, 1,4-Dihydroxyanthrahydroquinone, 1,4,9,10-Tetrahydroxyanthracene, 2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone

A745983-100G A745983-1G A745983-250MG A745983-25G
77199-178EA 133.9 USD
77199-178 77199-180 77199-182 77199-184
Leucoquinizarin 96%
Leucoquinizarin

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
MDL Number: MFCD00058948
CAS Number: 476-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR