1-Benzyl-1H-pyrazole-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Benzyl-1H-pyrazol-4-yl)boronic acid

A745665-100MG A745665-1G A745665-5G
77199-174EA 15 USD
77199-174 77199-176 77618-054
1-Benzyl-1H-pyrazole-4-boronic acid 98%
1-Benzyl-1H-pyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BN₂O₂
MDL Number: MFCD04038760
CAS Number: 852362-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR