4-Bromo-1-tosyl-1H-pyrazole 98%
Supplier: Ambeed
|
Danger
|
A743356-100MG
A743356-1G
A743356-250MG
A743356-5G
77170-988EA
14
USD
77170-988
77170-990
77170-992
77170-994
4-Bromo-1-tosyl-1H-pyrazole 98%
4-Bromo-1-tosyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₁₀H₉BrN₂O₂S |
MDL Number:
MFCD06080375 CAS Number: 116228-41-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...