2-(4-Nitrophenyl)propan-2-ol 95%

Supplier: Ambeed
Warning

A743201-100MG A743201-250MG A743201-1G
77170-984EA 77 USD
77170-984 77170-986 77893-471
2-(4-Nitrophenyl)propan-2-ol 95%
2-(4-Nitrophenyl)propan-2-ol
Formula: C₉H₁₁NO₃
MDL Number: MFCD00466951
CAS Number: 22357-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR