4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dibromo-5,6-difluorobenzo[c][1,2,5]thiadiazole

A741193-100MG A741193-250MG A741193-25G A741193-1G
77170-980EA 8.24 USD
77170-980 77170-982 77573-372 77595-166
4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole 97%
4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₆Br₂F₂N₂S
MW: 329.94 g/mol
MDL Number: MFCD28369502
CAS Number: 1295502-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR