Ethyl 5-methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A741174-100MG A741174-1G A741174-250MG
77170-974EA 111.24 USD
77170-974 77170-976 77170-978
Ethyl 5-methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%
Ethyl 5-methoxy-1H-pyrrolo[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃
MW: 220.22 g/mol
MDL Number: MFCD09833400
CAS Number: 3469-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR