1-Ethyl-1H-pyrazole-4-carbonitrile 97%

Supplier: Ambeed
Warning

A740803-1G A740803-250MG
77199-156EA 71.07 USD
77199-156 77199-158
1-Ethyl-1H-pyrazole-4-carbonitrile 97%
1-Ethyl-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₇N₃
MDL Number: MFCD04969705
CAS Number: 1006434-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR