Methyl-5-amino-2-fluorobenzenecarboxylate 98%

Supplier: Ambeed
Warning

A740780-10G A740780-250MG
77170-966EA 64 USD
77170-966 77170-968
Methyl-5-amino-2-fluorobenzenecarboxylate 98%
Methyl-5-amino-2-fluorobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD09037918
CAS Number: 56741-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR