3-(Trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine 98%

Supplier: Ambeed
Warning

A740768-100MG A740768-1G A740768-250MG
77199-150EA 49.44 USD
77199-150 77199-152 77199-154
3-(Trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine 98%
3-(Trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄F₃N₃
MW: 187.12 g/mol
MDL Number: MFCD09863958
CAS Number: 230305-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR