2-(1-Piperidinyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 3-Piperidinophenylboronic acid pinacol ester

A738778-1G A738778-250MG A738778-5G
77199-144EA 85.49 USD
77199-144 77199-146 77199-148
2-(1-Piperidinyl)phenylboronic acid pinacol ester 97%
2-(1-Piperidinyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₂
MW: 287.2100 g/mol
MDL Number: MFCD07368528
CAS Number: 852227-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR