2,4,6-Tri-p-tolylpyrylium tetrafluoroborate 98%

Supplier: Ambeed
Warning

A737649-100MG A737649-250MG A737649-50MG A737649-5G
77199-122EA 10.3 USD
77199-122 77199-124 77199-126 77747-459
2,4,6-Tri-p-tolylpyrylium tetrafluoroborate 98%
2,4,6-Tri-p-tolylpyrylium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₆H₂₃BF₄O
MW: 438.26 g/mol
MDL Number: MFCD00972530
CAS Number: 2713-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR