5-Bromo-2-iodo-4-methylaniline 96%

Supplier: Ambeed
Warning

A736959-100MG A736959-250MG
77199-118EA 33 USD
77199-118 77199-120
5-Bromo-2-iodo-4-methylaniline 96%
5-Bromo-2-iodo-4-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrIN
MDL Number: MFCD29037530
CAS Number: 1643156-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR