1-(4-(Trifluoromethyl)phenyl)cyclopropanamine 97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A736745-250MG
A736745-1G
77170-952EA
72.1
USD
77170-952
77684-661
1-(4-(Trifluoromethyl)phenyl)cyclopropanamine 97%, Ambeed.Inc
1-(4-(Trifluoromethyl)phenyl)cyclopropanamine
Precaución: For research use only.
Formula:
C₁₀H₁₀F₃N MW: 201.19 g/mol |
MDL Number:
MFCD07374475 CAS Number: 474709-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...