3-Chloro-5,5-dimethyl-2-cyclohexen-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5,5-dimethylcyclohex-2-enone

A735091-1G A735091-25G A735091-5G
77199-096EA 20.6 USD
77199-096 77199-098 77199-100
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one 98%
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₈H₁₁ClO
MW: 158.63 g/mol
MDL Number: MFCD00051620
CAS Number: 17530-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR