1,1,1-Tris(2-cyanoethyl)nitromethane 97%

Supplier: Ambeed
Warning

A734846-1G A734846-250MG
77199-092EA 20.6 USD
77199-092 77199-094
1,1,1-Tris(2-cyanoethyl)nitromethane 97%
1,1,1-Tris(2-cyanoethyl)nitromethane

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₂
MW: 220.22 g/mol
MDL Number: MFCD00013830
CAS Number: 1466-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR