1,1-Bis(methylthio)-2-nitroethylene 98%

Supplier: Ambeed
Danger

A733246-5G A733246-25G
77199-088EA 9 USD
77199-088 77617-984
1,1-Bis(methylthio)-2-nitroethylene 98%
1,1-Bis(methylthio)-2-nitroethylene

Precaución: for Research Use Only

Formula: C₄H₇NO₂S₂
MDL Number: MFCD00010443
CAS Number: 13623-94-4
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR