2-(2,2-Difluorocyclopropyl)acetic acid 98%

Supplier: Ambeed
Danger

A732873-250MG A732873-5G
77199-084EA 39 USD
77199-084 77199-086
2-(2,2-Difluorocyclopropyl)acetic acid 98%
2-(2,2-Difluorocyclopropyl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₆F₂O₂
MDL Number: MFCD19229279
CAS Number: 123131-67-9
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR