5-Bromo-2,4-difluorobenzamide 98+%

Supplier: Ambeed
Warning

A732682-10G A732682-250MG A732682-5G
77199-078EA 113 USD
77199-078 77199-080 77199-082
5-Bromo-2,4-difluorobenzamide 98+%
5-Bromo-2,4-difluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄BrF₂NO
MDL Number: MFCD28737880
CAS Number: 1805583-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR