(Bromomethyl)cyclopropane 97%

Supplier: Ambeed
Danger

A732217-10G A732217-1G A732217-25G A732217-5G
77170-936EA 9.27 USD
77170-936 77170-938 77170-940 77170-942
(Bromomethyl)cyclopropane 97%
(Bromomethyl)cyclopropane

Precaución: for Research Use Only

Formula: C₄H₇Br
MW: 135 g/mol
MDL Number: MFCD00001306
CAS Number: 7051-34-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR