2-Chloro-4,6-dimethyl-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A731987-100MG A731987-1G A731987-250MG
77199-072EA 54.59 USD
77199-072 77199-074 77199-076
2-Chloro-4,6-dimethyl-1,3,5-triazine 97%
2-Chloro-4,6-dimethyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
MDL Number: MFCD28671037
CAS Number: 30894-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR