5-Bromo-2-(3-fluorophenoxy)pyridine 97%

Supplier: Ambeed
Warning

A731061-1G A731061-250MG A731061-5G
77199-062EA 217 USD
77199-062 77199-064 77199-066
5-Bromo-2-(3-fluorophenoxy)pyridine 97%
5-Bromo-2-(3-fluorophenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇BrFNO
MDL Number: MFCD14652062
CAS Number: 936343-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR