b-Amino-4-bromobenzeneethanol 95%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: 2-Amino-2-(4-bromophenyl)ethanol

A730267-100MG A730267-1G A730267-5G
77170-922EA 39.14 USD
77170-922 77170-924 77684-646
b-Amino-4-bromobenzeneethanol 95%, Ambeed.Inc
b-Amino-4-bromobenzeneethanol

Precaución: For research use only.

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
MDL Number: MFCD12782005
CAS Number: 201403-02-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR