Methyl 4-(1,3-dioxan-2-yl)benzoate 97%

Supplier: Ambeed
Warning

A729181-1G A729181-250MG
77199-058EA 111.24 USD
77199-058 77199-060
Methyl 4-(1,3-dioxan-2-yl)benzoate 97%
Methyl 4-(1,3-dioxan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.23 g/mol
MDL Number: MFCD02612723
CAS Number: 135655-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR