1,3-Bis(4-chlorophenyl)thiourea 95+%

Supplier: Ambeed
Danger

A728176-1G A728176-250MG A728176-25G A728176-5G
77170-912EA 40.17 USD
77170-912 77170-914 77170-916 77170-918
1,3-Bis(4-chlorophenyl)thiourea 95+%
1,3-Bis(4-chlorophenyl)thiourea

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂N₂S
MW: 297.20 g/mol
MDL Number: MFCD00275761
CAS Number: 1220-00-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR