1,4-Butanediol diacrylate 90% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A726422-25G A726422-5G
77199-042EA 11.33 USD
77199-042 77199-044
1,4-Butanediol diacrylate 90% stabilized
1,4-Butanediol diacrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00014940
CAS Number: 1070-70-8
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR