7-Quinolinecarboxaldehyde 98%

Supplier: Ambeed
Warning

A726349-100MG A726349-1G A726349-250MG A726349-5G
77170-902EA 24 USD
77170-902 77170-904 77170-906 77170-908
7-Quinolinecarboxaldehyde 98%
7-Quinolinecarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MDL Number: MFCD01366121
CAS Number: 49573-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR