Methyl 4,5-dichloropicolinate 95%

Supplier: Ambeed
Warning

A724837-100MG A724837-250MG
77199-034EA 173.04 USD
77199-034 77199-036
Methyl 4,5-dichloropicolinate 95%
Methyl 4,5-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.02 g/mol
MDL Number: MFCD18257882
CAS Number: 1256834-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR