4-(4-Methoxyphenyl)pyridine 97%

Supplier: Ambeed
Warning

A724369-100MG A724369-1G A724369-250MG
77170-896EA 15 USD
77170-896 77170-898 77170-900
4-(4-Methoxyphenyl)pyridine 97%
4-(4-Methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MDL Number: MFCD00092331
CAS Number: 5938-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR