1,3,6,8-Tetraphenylpyrene 97%

Supplier: Ambeed
Warning

A724190-100MG A724190-250MG
77199-022EA 50.47 USD
77199-022 77199-024
1,3,6,8-Tetraphenylpyrene 97%
1,3,6,8-Tetraphenylpyrene

Precaución: for Research Use Only

Formula: C₄₀H₂₆
MW: 506.65 g/mol
MDL Number: MFCD00021662
CAS Number: 13638-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR