N-(3-Oxo-3-(4-(pyridin-4-yl)piperazin-1-yl)propyl)benzo[c][1,2,5]thiadiazole-4-sulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: VU0255035

A723827-100MG A723827-1MG A723827-25MG A723827-1G A723827-250MG
77170-890EA 49.44 USD
77170-890 77170-892 77170-894 77684-639 77684-640
N-(3-Oxo-3-(4-(pyridin-4-yl)piperazin-1-yl)propyl)benzo[c][1,2,5]thiadiazole-4-sulfonamide 95%
N-(3-Oxo-3-(4-(pyridin-4-yl)piperazin-1-yl)propyl)benzo[c][1,2,5]thiadiazole-4-sulfonamide

Precaución: For research use only.

Formula: C₁₈H₂₀N₆O₃S₂
MW: 432.52 g/mol
MDL Number: MFCD18086889
CAS Number: 1135243-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR