1-Benzylpyrrolidin-3-ylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Phenylmethyl)-3-pyrrolidinemethanol

A723241-1G A723241-25G A723241-5G
77170-884EA 23.69 USD
77170-884 77170-886 77170-888
1-Benzylpyrrolidin-3-ylmethanol 98%
1-Benzylpyrrolidin-3-ylmethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
MW: 191.28 g/mol
MDL Number: MFCD04038527
CAS Number: 5731-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR