1-Benzyl-3,3-difluoro-4,4-piperidinediol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3,3-difluoropiperidine-4,4-diol

A723222-250MG A723222-5G
77170-880EA 22.66 USD
77170-880 77170-882
1-Benzyl-3,3-difluoro-4,4-piperidinediol 98%
1-Benzyl-3,3-difluoro-4,4-piperidinediol

Precaución: for Research Use Only

Formula: C₁₂H₁₅F₂NO₂
MW: 243.26 g/mol
MDL Number: MFCD20926176
CAS Number: 1067914-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR