4-(2-Methoxy-benzyl)-piperidine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Methoxybenzyl)piperidine hydrochloride

A722733-1G A722733-250MG
77199-016EA 115 USD
77199-016 77199-018
4-(2-Methoxy-benzyl)-piperidine hydrochloride 95%
4-(2-Methoxy-benzyl)-piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO·ClH
MDL Number: MFCD08437508
CAS Number: 37581-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR