tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate 98%

Supplier: Ambeed
Danger

Synonyms: 4-(4-BOC-Pyrazino)benzylamine, 4-[(Aminomethyl)phenyl]piperazine, N1-BOC protected, tert-Butyl 4-(4-(aminomethyl)phenyl)piperazine-1-carboxylate

A722681-100MG A722681-1G A722681-250MG
77170-874EA 43 USD
77170-874 77170-876 77170-878
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate 98%
tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₅N₃O₂
MDL Number: MFCD03791213
CAS Number: 852180-47-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR