1-(4-Bromophenyl)cyclopropanamine hydrochloride 95%

Supplier: Ambeed
Warning

A722650-1G A722650-250MG A722650-5G
77170-868EA 62 USD
77170-868 77170-870 77170-872
1-(4-Bromophenyl)cyclopropanamine hydrochloride 95%
1-(4-Bromophenyl)cyclopropanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MDL Number: MFCD13195821
CAS Number: 952289-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR