4-Bromo-2,6-diethylbenzonitrile 95+%

Supplier: Ambeed
Warning

A722142-1G A722142-250MG A722142-5G
77170-862EA 39 USD
77170-862 77170-864 77170-866
4-Bromo-2,6-diethylbenzonitrile 95+%
4-Bromo-2,6-diethylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrN
MDL Number: MFCD18917122
CAS Number: 1200131-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR