7-Methoxy-1,1-dimethyl-3,4-dihydronaphthalen-2(1H)-one 98%

Supplier: Ambeed
Warning

A721590-100MG A721590-1G A721590-250MG
77170-856EA 39.14 USD
77170-856 77170-858 77170-860
7-Methoxy-1,1-dimethyl-3,4-dihydronaphthalen-2(1H)-one 98%
7-Methoxy-1,1-dimethyl-3,4-dihydronaphthalen-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MW: 204.26 g/mol
MDL Number: MFCD20274514
CAS Number: 1865-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR