2-Amino-4-chloro-3-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A721076-100MG A721076-1G A721076-250MG A721076-5G
77170-848EA 14 USD
77170-848 77170-850 77170-852 77170-854
2-Amino-4-chloro-3-methylbenzoic acid 95%
2-Amino-4-chloro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD01925304
CAS Number: 98968-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR