Phenyl(pyridin-4-yl)methanamine 95%

Supplier: Ambeed
Warning

A720889-100MG A720889-1G A720889-250MG
77170-842EA 31 USD
77170-842 77170-844 77170-846
Phenyl(pyridin-4-yl)methanamine 95%
Phenyl(pyridin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MDL Number: MFCD06589768
CAS Number: 58088-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR