6-Chloro-3-fluoropyridin-2-ol 95%

Supplier: Ambeed
Warning

A720208-100MG A720208-250MG A720208-1G
77170-836EA 37 USD
77170-836 77170-838 77552-230
6-Chloro-3-fluoropyridin-2-ol 95%
6-Chloro-3-fluoropyridin-2-ol

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
MDL Number: MFCD22547061
CAS Number: 1369769-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR