Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Supplier: Ambeed
|
Warning
|
A720014-1G
A720014-250MG
A720014-5G
77199-000EA
195.7
USD
77199-000
77199-002
77199-004
Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BO₅ MW: 292.14 g/mol |
MDL Number:
MFCD22494011 CAS Number: 478375-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...