2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A719022-100MG A719022-250MG A719022-1G
77170-832EA 39 USD
77170-832 77170-834 77573-322
2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine 98%
2,4-Dichloro-6-methylthieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂S
MDL Number: MFCD15144570
CAS Number: 76872-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR