Methyl 4-amino-3,5-difluorobenzoate 95%

Supplier: Ambeed
Warning

A719008-100MG A719008-1G A719008-250MG A719008-5G
77170-826EA 11 USD
77170-826 77170-828 77170-830 77893-435
Methyl 4-amino-3,5-difluorobenzoate 95%
Methyl 4-amino-3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MDL Number: MFCD20725083
CAS Number: 1162676-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR