Benzo[b]thiophen-4-amine 98%

Supplier: Ambeed
Warning

A718407-100MG A718407-1G A718407-250MG A718407-5G
77198-990EA 90.64 USD
77198-990 77198-992 77198-994 77198-996
Benzo[b]thiophen-4-amine 98%
Benzo[b]thiophen-4-amine

Precaución: for Research Use Only

Formula: C₈H₇NS
MW: 149.21 g/mol
MDL Number: MFCD01860288
CAS Number: 17402-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR