1,4-Dimethoxynaphthalene 98%

Supplier: Ambeed
Warning

A718100-10G A718100-1G A718100-250MG A718100-5G
77198-982EA 372.86 USD
77198-982 77198-984 77198-986 77198-988
1,4-Dimethoxynaphthalene 98%
1,4-Dimethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
MDL Number: MFCD00052378
CAS Number: 10075-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR