(5-Chloro-1H-indol-2-yl)methanol 98+%

Supplier: Ambeed
Warning

A716014-100MG A716014-1G A716014-250MG
77170-812EA 52.53 USD
77170-812 77170-814 77170-816
(5-Chloro-1H-indol-2-yl)methanol 98+%
(5-Chloro-1H-indol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.619 g/mol
MDL Number: MFCD19301593
CAS Number: 53590-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR